CAS RN 2156588-26-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ ARCC 4 negative control
Negative control for ARCC 4 (Cat. No. 7254)