accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21587-74-6

IUPAC Name: 3-[9-(3-aminopropyl)-2,4,8,10-tetraoxaspiro[5.5]undecan-3-yl]propan-1-amine
Synonyms: 3-[9-(3-aminopropyl)-2,4,8,10-tetraoxaspiro[5.5]undecan-3-yl]propan-1-amine
Molecular Weight (g/mol): 274.36
Molecular Formula: C13H26N2O4
InChi Key: ANOPCGQVRXJHHD-UHFFFAOYSA-N
SMILES: NCCCC1OCC2(CO1)COC(CCCN)OC2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results