accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOO

CAS RN 215923-54-9

IUPAC Name: 1-(2,3-dihydro-1,4-benzodioxine-6-carbonyl)piperidine
Synonyms: 1-(2,3-dihydro-1,4-benzodioxine-6-carbonyl)piperidine
Molecular Weight (g/mol): 247.29
Molecular Formula: C14H17NO3
InChi Key: LJUNPHMOGNFFOS-UHFFFAOYSA-N
SMILES: O=C(N1CCCCC1)C1=CC2=C(OCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)