accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2162-99-4

IUPAC Name: 1,8-dichlorooctane
Synonyms: octane, 1,8-dichloro-
Molecular Weight (g/mol): 183.12
Molecular Formula: C8H16Cl2
InChi Key: WXYMNDFVLNUAIA-UHFFFAOYSA-N
SMILES: ClCCCCCCCCCl
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)