Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Dichlorooctane 99.0+%, TCI America™
SDP

Catalog No. D101225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D101225G 25 g
D101210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D101225G Supplier TCI America Supplier No. D101225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2162-99-4
Molecular Formula C8H16Cl2
Molecular Weight (g/mol) 183.12
MDL Number MFCD00039393
InChI Key WXYMNDFVLNUAIA-UHFFFAOYSA-N
Synonym octane, 1,8-dichloro, 1,8-dichloroethane, octamethylene chloride, octamethylene dichloride, acmc-209fmk, 1,8-dichlorooctane
PubChem CID 75102
IUPAC Name 1,8-dichlorooctane
SMILES ClCCCCCCCCCl

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 g
UN Number 3082
Formula Weight 183.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,8-Dichlorooctane
EINECSNumber (2)-0066
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.