accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOO

CAS RN 2164-34-3

IUPAC Name: 2-(bromomethyl)-2,3-dihydro-1,4-benzodioxine
Synonyms: 2-(bromomethyl)-2,3-dihydro-1,4-benzodioxine
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: QYLFKNVZIFTCIY-UHFFFAOYNA-N
SMILES: BrCC1COC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results