accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrOH

CAS RN 216755-56-5

IUPAC Name: (3-bromo-5-fluorophenyl)methanol
Synonyms: (3-bromo-5-fluorophenyl)methanol
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: KQWBQTOZXPPFIY-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results