Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-fluorobenzyl alcohol, 98%, Thermo Scientific™

Catalog No. AAH61415MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61415MD 250 mg
AAH6141503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61415MD Supplier Thermo Scientific Chemicals Supplier No. H61415MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216755-56-5
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD04116320
InChI Key KQWBQTOZXPPFIY-UHFFFAOYSA-N
Synonym 3-bromo-5-fluorobenzyl alcohol, 3-bromo-5-fluorophenyl methanol, 3-bromo-5-fluorobenzylalcohol, 5-bromo-3-fluorobenzyl alcohol, benzenemethanol,3-bromo-5-fluoro, benzenemethanol, 3-bromo-5-fluoro, 3-bromo-5-fluorophenyl methan-1-ol, 3-bromo-5-fluoro-phenyl methanol, pubchem4885, acmc-1cgon
PubChem CID 2769402
IUPAC Name (3-bromo-5-fluorophenyl)methanol
SMILES C1=C(C=C(C=C1F)Br)CO

Specifications

Color Colorless
Quantity 250 mg
Formula Weight 205.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-fluorobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.