CAS RN 2172820-12-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ VH 032 amide-PEG4-acid
VHL ligand with PEG linker and terminal acid for onward chemistry