accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SnH3CH3CH3C

CAS RN 2176-98-9

IUPAC Name: tetrapropylstannane
Synonyms: tetrapropyl tin
Molecular Weight (g/mol): 291.07
Molecular Formula: C12H28Sn
InChi Key: OIQCWAIEHVRCCG-UHFFFAOYSA-N
SMILES: CCC[Sn](CCC)(CCC)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results