accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21794-01-4

IUPAC Name: 2-hydroxy-10-[hydroxy(6-oxo-3,6-dihydro-2H-pyran-2-yl)methyl]-3-(1-hydroxyheptyl)-6,14-dioxatricyclo[10.3.0.0⁴,⁸]pentadeca-1(12),4(8)-diene-5,7,13,15-tetrone
Synonyms: rubratoxin B
Molecular Weight (g/mol): 518.52
Molecular Formula: C26H30O11
InChi Key: ZJTBTDVZNGBSNG-UHFFFAOYNA-N
SMILES: CCCCCCC(O)C1C(O)C2=C(CC(CC3=C1C(=O)OC3=O)C(O)C1CC=CC(=O)O1)C(=O)OC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results