accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNHNN

CAS RN 21871-47-6

IUPAC Name: 5-(benzylsulfanyl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(benzylsulfanyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 192.24
Molecular Formula: C8H8N4S
InChi Key: GXGKKIPUFAHZIZ-UHFFFAOYSA-N
SMILES: C(SC1=NNN=N1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)