CAS RN 2189-27-7
CAS RN 2189-27-7
IUPAC Name:
2-(2-aminoacetamido)pentanoic acid
Synonyms:
2-(2-aminoacetamido)pentanoic acid
Molecular Weight (g/mol):
174.2
Molecular Formula:
C7H14N2O3
InChi Key:
JXIQKLAZYWZTRA-UHFFFAOYNA-N
SMILES:
CCCC(NC(=O)CN)C(O)=O
1
–
1
of
1
results
1
Glycyl-DL-norvaline 97.0+%, TCI America™
CAS: 2189-27-7 Molecular Formula: C7H14N2O3 Molecular Weight (g/mol): 174.2 MDL Number: MFCD00038186 InChI Key: JXIQKLAZYWZTRA-UHFFFAOYSA-N Synonym: H-Gly-DL-Nva-OH PubChem CID: 86841 IUPAC Name: 2-[(2-aminoacetyl)amino]pentanoic acid SMILES: CCCC(C(=O)O)NC(=O)CN