accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21906-39-8

IUPAC Name: 1-[3-(trifluoromethyl)phenyl]propan-2-one
Synonyms: 1-[3-(trifluoromethyl)phenyl]propan-2-one
Molecular Weight (g/mol): 202.18
Molecular Formula: C10H9F3O
InChi Key: JPHQCDCEBDRIOL-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=CC(=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results