accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOFFFFF

CAS RN 2192-55-4

IUPAC Name: 2-(2,3,4,5,6-pentafluorophenoxy)ethan-1-ol
Synonyms: 2-(2,3,4,5,6-pentafluorophenoxy)ethanol
Molecular Weight (g/mol): 228.12
Molecular Formula: C8H5F5O2
InChi Key: NPVVUYMVRISVGK-UHFFFAOYSA-N
SMILES: OCCOC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results