CAS RN 219832-49-2
CAS RN 219832-49-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ MP A08
Selective ATP competitive sphingosine kinase 2 and 1 (SphK2/1) inhibitor
2
Tocris Bioscience™ MP A08
Selective ATP competitive SphK1 and 2 inhibitor; cell permeable