accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3H3CH3CNOH(E)

CAS RN 22033-87-0

IUPAC Name: N-[(7E)-9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ylidene]hydroxylamine
Synonyms: olesoxime
Molecular Weight (g/mol): 399.66
Molecular Formula: C27H45NO
InChi Key: QNTASHOAVRSLMD-SGWCAAJKNA-N
SMILES: CC(C)CCCC(C)C1CCC2C3CCC4=C\C(CCC4(C)C3CCC12C)=N\O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)