accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 220662-95-3

IUPAC Name: methyl[4-(pyridin-3-yl)but-3-en-1-yl]amine; oxalic acid
Synonyms: oxalic acid; rivanicline
Molecular Weight (g/mol): 252.27
Molecular Formula: C12H16N2O4
InChi Key: WTIZFOAIQXMQHC-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.CNCCC=CC1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results