CAS RN 2211-98-5
IUPAC Name:
sodium 4-dodecylbenzene-1-sulfonate
Synonyms:
dodecylbenzenesulfonic acid, NA salt
Molecular Weight (g/mol):
348.48
Molecular Formula:
C18H29NaO3S
InChi Key:
JHJUUEHSAZXEEO-UHFFFAOYSA-M
SMILES:
[Na+].CCCCCCCCCCCCC1=CC=C(C=C1)S([O-])(=O)=O
1
–
1
of
1
results
1
Dodecylbenzene Sodium Sulfonate, 65% in water, MP Biomedicals
CAS: 2211-98-5 Molecular Formula: C18H29NaO3S Molecular Weight (g/mol): 348.477 InChI Key: JHJUUEHSAZXEEO-UHFFFAOYSA-M Synonym: sodium 4-dodecylbenzenesulfonate,sodium p-dodecylbenzenesulfonate,conoco c-50,unii-hb2d2zei04,hb2d2zei04,benzenesulfonic acid, 4-dodecyl-, sodium salt 1:1,dodecene-1 las,benzenesulfonic acid, 4-dodecyl-, sodium salt,sodium 4-dodecylbenzenesulphonate,4-dodecylbenzenesulfonic acid sodium PubChem CID: 4289524 IUPAC Name: sodium;4-dodecylbenzenesulfonate SMILES: CCCCCCCCCCCCC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]