Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylbenzene Sodium Sulfonate, 65% in water, MP Biomedicals

Catalog No. ICN203311 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203311 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203311 Supplier MP Biomedicals, Inc Supplier No. 0520331125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2211-98-5
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.477
InChI Key JHJUUEHSAZXEEO-UHFFFAOYSA-M
Synonym sodium 4-dodecylbenzenesulfonate, sodium p-dodecylbenzenesulfonate, conoco c-50, unii-hb2d2zei04, hb2d2zei04, benzenesulfonic acid, 4-dodecyl-, sodium salt 1:1, dodecene-1 las, benzenesulfonic acid, 4-dodecyl-, sodium salt, sodium 4-dodecylbenzenesulphonate, 4-dodecylbenzenesulfonic acid sodium
PubChem CID 4289524
IUPAC Name sodium;4-dodecylbenzenesulfonate
SMILES CCCCCCCCCCCCC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]

Specifications

Color Cream-White
Quantity 25 g
Formula Weight 348.49
Physical Form Paste
Chemical Name or Material Dodecylbenzene Sodium Sulfonate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.