accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2217-07-4

IUPAC Name: N,N-dipropylaniline
Synonyms: benzenamine, N,N-dipropyl-
Molecular Weight (g/mol): 177.29
Molecular Formula: C12H19N
InChi Key: MMFBQHXDINNBMW-UHFFFAOYSA-N
SMILES: CCCN(CCC)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)