Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dipropylaniline 95%, Thermo Scientific™

Catalog No. AC409140250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC409140250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC409140250 Supplier Thermo Scientific Chemicals Supplier No. 409140250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2217-07-4
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.29
InChI Key MMFBQHXDINNBMW-UHFFFAOYSA-N
Synonym n,n-di-n-propylaniline, benzenamine, n,n-dipropyl, dipropylaniline, dipropylamine, n-phenyl, n-phenyldipropylamine, unii-bz9f2k19ke, bz9f2k19ke, n,n-di-n-propyl benzenamine, phenyldipropylamine, aniline,n-dipropyl
PubChem CID 75191
IUPAC Name N,N-dipropylaniline
SMILES CCCN(CCC)C1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown
Density 0.91
Infrared Spectrum Authentic
Refractive Index 1.5250 to 1.5290 (20°C, 589nm)
Quantity 25 g
Formula Weight 177.29
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material N, N-Dipropylaniline, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.