Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dipropylaniline 98.0+%, TCI America™
SDP

Catalog No. D194125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D194125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D194125ML Supplier TCI America Supplier No. D194125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2217-07-4
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.29
MDL Number MFCD00035797
InChI Key MMFBQHXDINNBMW-UHFFFAOYSA-N
Synonym n,n-di-n-propylaniline, benzenamine, n,n-dipropyl, dipropylaniline, dipropylamine, n-phenyl, n-phenyldipropylamine, unii-bz9f2k19ke, bz9f2k19ke, n,n-di-n-propyl benzenamine, phenyldipropylamine, aniline,n-dipropyl
PubChem CID 75191
IUPAC Name N,N-dipropylaniline
SMILES CCCN(CCC)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 mL
Formula Weight 177.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Dipropylaniline
EINECSNumber (3)-0129
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.