accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3OHO

CAS RN 22204-53-1

IUPAC Name: 2-(6-methoxynaphthalen-2-yl)propanoic acid
Synonyms: naprosyn
Molecular Weight (g/mol): 230.26
Molecular Formula: C14H14O3
InChi Key: CMWTZPSULFXXJA-UHFFFAOYNA-N
SMILES: COC1=CC=C2C=C(C=CC2=C1)C(C)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (9)
Quantity 
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)
  • (3)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (9)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)