Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naproxen, MP Biomedicals

Catalog No. ICN19024750 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19024750 50 g
ICN19024701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19024750 Supplier MP Biomedicals, Inc Supplier No. 0219024750
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22204-53-1
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.263
InChI Key CMWTZPSULFXXJA-VIFPVBQESA-N
Synonym naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno
PubChem CID 156391
ChEBI CHEBI:7476
IUPAC Name (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.