accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22245-83-6

IUPAC Name: 3-(trifluoromethyl)-1,2-dihydropyridin-2-one
Synonyms: 3-(trifluoromethyl)-1H-pyridin-2-one
Molecular Weight (g/mol): 163.1
Molecular Formula: C6H4F3NO
InChi Key: JHDCDEHVUADNKQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CNC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results