CAS RN 2228029-82-9
CAS RN 2228029-82-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Pomalidomide 5'-piperazine
Cereblon ligand with terminal piperazine for onward chemistry