CAS RN 2230613-03-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Ortho AP 1867
Selective binding ligand for FKBP12F36V; precursor for dTAG compounds