accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NOHO

CAS RN 22426-30-8

IUPAC Name: 2-cyano-2,2-dimethylacetic acid
Synonyms: 2-cyano-2,2-dimethylacetic acid
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: DZDVNAVPCCNJRK-UHFFFAOYSA-N
SMILES: CC(C)(C#N)C(O)=O
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results