accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22451-06-5

H2OH2OH2OH2OH2ONaNaOTeOOO

CAS RN 22451-06-5

IUPAC Name: disodium pentahydrate tellurate
Synonyms: sodium tellurite
Molecular Weight (g/mol): 327.65
Molecular Formula: H10Na2O9Te
InChi Key: OVBVNEXDNNHXIU-UHFFFAOYSA-L
SMILES: O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)