accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2246-46-0

IUPAC Name: (4E)-3-hydroxy-4-[2-(1,3-thiazol-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Synonyms: (4E)-3-hydroxy-4-[2-(1,3-thiazol-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 221.23
Molecular Formula: C9H7N3O2S
InChi Key: UVQWEDPLIFNWSH-YRNVUSSQSA-N
SMILES: OC1=CC(=O)C=C\C1=N/NC1=NC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results