Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAR 99.0+%, TCI America™
SDP

Catalog No. A50025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50025G 5 g
A50021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50025G Supplier TCI America Supplier No. A50025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[Metal indicator and spectrophotometric reagent for transition metals]

Chemical Identifiers

CAS 2246-46-0
Molecular Formula C9H7N3O2S
Molecular Weight (g/mol) 221.23
MDL Number MFCD00005322
InChI Key UVQWEDPLIFNWSH-YRNVUSSQSA-N
Synonym 4-2-thiazolylazo resorcinol, 1,3-benzenediol, 4-2-thiazolylazo, 2-2,4-dihydroxyphenylazo thiazole, 4-thiazol-2-ylazo resorcinol, 4-thiazol-2-yldiazenyl benzene-1,3-diol, 4-2-1,3-thiazol-2-yl diazen-1-yl benzene-1,3-diol, 4-e-2-1,3-thiazol-2-yl diazen-1-yl benzene-1,3-diol, shnikuxmzfppcs-vawyxsnfsa-n, 4-e-1,3-thiazol-2-yldiazenyl-1,3-benzenediol #
PubChem CID 9571124
IUPAC Name (4E)-3-hydroxy-4-[2-(1,3-thiazol-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=C\C1=N/NC1=NC=CS1

Specifications

Melting Point 209°C
Quantity 5 g
Formula Weight 221.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material TAR [=4-(2-Thiazolylazo)resorcinol] [Metal indicator and spectrophotometric reagent for trans
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.