CAS RN 2250025-92-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ DREADD agonist 21 dihydrochloride
Potent muscarinic DREADD agonist; water soluble version of DREADD agonist 21 (Cat. No. 5548)