accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFClO

CAS RN 2251-50-5

IUPAC Name: pentafluorobenzoyl chloride
Synonyms: pentafluorobenzoyl chloride
Molecular Weight (g/mol): 230.52
Molecular Formula: C7ClF5O
InChi Key: MYHOHFDYWMPGJY-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C(Cl)=O)C(F)=C1F
Percent Purity 
  • (2)
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (4)
  • (4)
Boiling Point 
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (2)
  • (6)
  • (3)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (2)
  • (5)