accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22551-26-4

IUPAC Name: 3-(methylsulfanyl)propane-1,2-diol
Synonyms: 3-(methylsulfanyl)propane-1,2-diol
Molecular Weight (g/mol): 122.18
Molecular Formula: C4H10O2S
InChi Key: FXIHXPKPLPKPJH-UHFFFAOYNA-N
SMILES: CSCC(O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results