accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 225518-49-0

IUPAC Name: 5,7-dichloro-2H,3H-thieno[3,4-b][1,4]dioxine
Synonyms: 5,7-dichloro-2H,3H-thieno[3,4-b][1,4]dioxine
Molecular Weight (g/mol): 211.06
Molecular Formula: C6H4Cl2O2S
InChi Key: UOJXEABHQWNIKN-UHFFFAOYSA-N
SMILES: ClC1=C2OCCOC2=C(Cl)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results