Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloro-3,4-ethylenedioxythiophene 98.0+%, TCI America™
SDP

Catalog No. D47461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D47461G Supplier TCI America Supplier No. D47461G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 225518-49-0
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.056
MDL Number MFCD09907757
InChI Key UOJXEABHQWNIKN-UHFFFAOYSA-N
Synonym 5,7-Dichloro-2,3-dihydrothieno[3,4-b][1,4]dioxine
PubChem CID 22177487
IUPAC Name 5,7-dichloro-2,3-dihydrothieno[3,4-b][1,4]dioxine
SMILES C1COC2=C(SC(=C2O1)Cl)Cl

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 1 g
Formula Weight 211.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloro-3,4-ethylenedioxythiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.