accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22577-15-7

IUPAC Name: 3-(prop-2-en-1-yloxy)propanoic acid
Synonyms: 3-(prop-2-en-1-yloxy)propanoic acid
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: LBJZZFXUVYHXPH-UHFFFAOYSA-N
SMILES: OC(=O)CCOCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results