Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Allyloxypropionic Acid 95.0+%, TCI America™
SDP

Catalog No. A065710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A065710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A065710G Supplier TCI America Supplier No. A065710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22577-15-7
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00039537
InChI Key LBJZZFXUVYHXPH-UHFFFAOYSA-N
PubChem CID 552244
IUPAC Name 3-prop-2-enoxypropanoic acid
SMILES C=CCOCCC(=O)O

Specifications

Color Yellow
Boiling Point 192°C
Quantity 10 g
Formula Weight 130.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Allyloxypropionic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.