CAS RN 2259-96-3
IUPAC Name:
3-{bicyclo[2.2.1]hept-5-en-2-yl}-6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ⁶,2,4-benzothiadiazine-7-sulfonamide
Synonyms:
cyclothiazide fluidil renazide doburil cyclothiazide preparation cyclothiazide (substance) 环噻嗪 cyclothiazidum aquirel anhydron
Molecular Weight (g/mol):
389.87
Molecular Formula:
C14H16ClN3O4S2
InChi Key:
BOCUKUHCLICSIY-UHFFFAOYNA-N
SMILES:
NS(=O)(=O)C1=CC2=C(NC(NS2(=O)=O)C2CC3CC2C=C3)C=C1Cl