accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 226570-67-8

IUPAC Name: bromozincylium; but-3-en-1-ide
Synonyms: bromozincylium; but-3-en-1-ide
Molecular Weight (g/mol): 200.38
Molecular Formula: C4H7BrZn
InChi Key: BFFDFNINBXPQIU-UHFFFAOYSA-M
SMILES: [Zn+]Br.[CH2-]CC=C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)