accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(R)(S)H3C(S)(S)H3CO(S)O(R)H3C(S)

CAS RN 2268-98-6

IUPAC Name: (1S,3aS,3bS,9aR,9bS,10R,11aS)-1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-10-yl acetate
Synonyms: (1S,3aS,3bS,9aR,9bS,10R,11aS)-1-acetyl-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,8H,9H,9bH,10H,11H-cyclopenta[a]phenanthren-10-yl acetate
Molecular Weight (g/mol): 372.51
Molecular Formula: C23H32O4
InChi Key: IWRPVTXREVYBHT-ZQEATNLPSA-N
SMILES: CC(=O)O[C@@H]1C[C@@]2(C)[C@@H](CC[C@@H]2C(C)=O)[C@@H]2CCC3=CC(=O)CC[C@]3(C)[C@@H]12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results