accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOSNONOO

CAS RN 227088-94-0

IUPAC Name: 3-[1-({3'-nitro-[1,1'-biphenyl]-4-yl}oxy)-4-(pyridin-4-yl)butan-2-yl]-1,3-thiazolidine-2,4-dione
Synonyms: 3-[1-({3'-nitro-[1,1'-biphenyl]-4-yl}oxy)-4-(pyridin-4-yl)butan-2-yl]-1,3-thiazolidine-2,4-dione
Molecular Weight (g/mol): 463.51
Molecular Formula: C24H21N3O5S
InChi Key: VQEHBLGYANQWEA-UHFFFAOYNA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C1=CC=C(OCC(CCC2=CC=NC=C2)N2C(=O)CSC2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)