accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHO

CAS RN 2284-30-2

IUPAC Name: 4-benzylbenzene-1,3-diol
Synonyms: 4-benzylbenzene-1,3-diol
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: QVFIWTNWKHFVEH-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC2=CC=CC=C2)C(O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results