accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 22856-30-0

IUPAC Name: naphthalene-2,3-dicarbonitrile
Synonyms: naphthalene-2,3-dicarbonitrile
Molecular Weight (g/mol): 178.19
Molecular Formula: C12H6N2
InChi Key: KNBYJRSSFXTESR-UHFFFAOYSA-N
SMILES: N#CC1=C(C=C2C=CC=CC2=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results