accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22868-35-5

IUPAC Name: 2-(3-chlorophenyl)-1H-1,3-benzodiazole
Synonyms: 2-(3-chlorophenyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 228.68
Molecular Formula: C13H9ClN2
InChi Key: BGYHTIIVIGGCIF-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results