CAS RN 229005-80-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ TAK 779
Potent and selective CCR5 and CCR2 chemokine receptor antagonist