accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2295-35-4

IUPAC Name: pyrrolidine-2-thione
Synonyms: 2-azacyclopentanthione
Molecular Weight (g/mol): 101.17
Molecular Formula: C4H7NS
InChi Key: IMWUREPEYPRYOR-UHFFFAOYSA-N
SMILES: S=C1CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results