Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrolidine-2-thione, 97%, Thermo Scientific™

Catalog No. AAL0937406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0937406 5 g
AAL0937403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0937406 Supplier Thermo Scientific Chemicals Supplier No. L0937406

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2295-35-4
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD00049351
InChI Key IMWUREPEYPRYOR-UHFFFAOYSA-N
Synonym 2-pyrrolidinethione, cis-2-pyrrolidinethione, cis-tetrahydro-2-pyrrolethione, 2-azacyclopentanthione, 2-pyrrolethione, tetrahydro-, z, .gamma.-butyrothiolactam, thiopyrrolidone, thiobutyrolactam, pubchem20272, acmc-1cr4h
PubChem CID 2775267
IUPAC Name pyrrolidine-2-thione
SMILES C1CC(=S)NC1

Specifications

Melting Point 109°C to 113°C
Quantity 5 g
Beilstein 105242
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Pyrrolidine-2-thione
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UX9647500
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.