accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3NHOHOHHOH3CH3CCH3NHOHOHHOOHSOHOO

CAS RN 23031-32-5

IUPAC Name: bis(5-[2-(tert-butylamino)-1-hydroxyethyl]benzene-1,3-diol); sulfuric acid
Synonyms: terbutaline sulfate
Molecular Weight (g/mol): 548.65
Molecular Formula: C24H40N2O10S
InChi Key: KFVSLSTULZVNPG-UHFFFAOYNA-N
SMILES: OS(O)(=O)=O.CC(C)(C)NCC(O)C1=CC(O)=CC(O)=C1.CC(C)(C)NCC(O)C1=CC(O)=CC(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)